Learn More
Cayman Chemical THUNDERPhospho-ERK12T202 96Wel
SDP
Supplier:  Cayman Chemical 50020696 WELL
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-ERK1/2 (T202/Y204) in cell lysates.
Catalog No. 50-289-4082