Learn More
Cayman Chemical THUNDERPhospho-p38a MAPK 96Wel
SDP
Supplier:  Cayman Chemical 50026996 WELL
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-p38αβγ MAPK14/11/12 (T180/Y182) in cell lysates
Catalog No. 50-289-3993