Learn More
Cayman Chemical THUNDERPhospho-p53S15 TR 96Wel
SDP
Supplier:  Cayman Chemical 50027196 WELL
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-p53 (S15) in cell lysates
Catalog No. 50-289-4085