Learn More
Cayman Chemical THUNDERPhospho-STAT3Y705 96Wel
SDP
Supplier:  Cayman Chemical 50020096 WELL
Encompass_Preferred
A sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-STAT3 (Y705) in cell lysates
Catalog No. 50-289-4649