Learn More
Cayman Chemical THUNDERPhosphoGSK3S9 T 2400
SDP
Supplier:  Cayman Chemical 5002512400 WELL
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-GSK3β (S9) in cell lysates
Catalog No. 50-289-6550