Learn More
Cayman Chemical THUNDERPhosphoSTAT3Y705 480
SDP
Supplier:  Cayman Chemical 500200480 WELL
Encompass_Preferred
A sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-STAT3 (Y705) in cell lysates
Catalog No. 50-289-6549