Learn More
Cayman Chemical THUNDRcAMP TRFRET asy K 19200
SDP
Supplier:  Cayman Chemical 50029319200 WELL
Encompass_Preferred
A homogeneous competitive TR-FRET immunoassay for quantitative measurement of cAMP in cell lysates
Catalog No. 50-289-3362