Learn More
Cayman Chemical THUNDREXreme PhosphoRK1 2400
SDP
Supplier:  Cayman Chemical 5002912400 WELL
Encompass_Preferred
A sandwich TR-FRET immunoassay for semi-quantitative detection of low levels of Phospho-ERK1/2 (T202/Y204) in cell lysates
Catalog No. 50-289-4354