Learn More
Cayman Chemical THUNDRHumn CCL2MCP1 TRF 4800
SDP
Supplier:  Cayman Chemical 5002374800 WELL
Encompass_Preferred
Sandwich TR-FRET immunoassay for the quantitative measurement of human CCL2 (MCP-1; MCAF; JE) in cell culture supernatants
Catalog No. 50-289-5534