Learn More
Cayman Chemical THUNDRHumn IL18 TRFRET 9600Wel
SDP
Supplier:  Cayman Chemical 5003049600 WELL
Encompass_Preferred
A sandwich TR-FRET immunoassay for the quantitative measurement of human IL‑18 (IL-1F4) in cell culture supernatants
Catalog No. 50-289-5961
Only null left
Add to Cart
Request a Quote