Learn More
Cayman Chemical THUNDRHumn IL2 TRFRET B 480Wel
SDP
Supplier:  Cayman Chemical 500231480 WELL
Encompass_Preferred
Sandwich TR-FRET immunoassay for the quantitative measurement of human IL-2 in cell culture supernatants
Catalog No. 50-289-5200