Learn More
Cayman Chemical THUNDRHumn TNFa TRFRET B 480
SDP
Supplier:  Cayman Chemical 500234480 WELL
Encompass_Preferred
Sandwich TR-FRET immunoassay for the quantitative measurement of human TNFα in cell culture supernatants
Catalog No. 50-289-4212