Learn More
Cayman Chemical THUNDRMouse IL2 TRFRET B 480
SDP
Supplier:  Cayman Chemical 500311480 WELL
Encompass_Preferred
A sandwich TR-FRET immunoassay for the quantitative measurement of mouse IL-2 in cell culture supernatants
Catalog No. 50-289-5268