Learn More
Cayman Chemical THUNDRMouse IL2 TRFRET B 9600
SDP
Supplier:  Cayman Chemical 5003119600 WELL
Encompass_Preferred
A sandwich TR-FRET immunoassay for the quantitative measurement of mouse IL-2 in cell culture supernatants
Catalog No. 50-289-5842
Only null left
Add to Cart
Request a Quote