Learn More
Cayman Chemical THUNDRPhosphocMetY1234 2400Wel
SDP
Supplier:  Cayman Chemical 5002622400 WELL
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-c-Met (Y1234/Y1235) in cell lysates
Catalog No. 50-289-6058