Learn More
Cayman Chemical THUNDRPhosphoInsulIn Rece 96
SDP
Supplier:  Cayman Chemical 50020996 WELL
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-Insulin Receptor β (Y1150/Y1151) in cell lysates
Catalog No. 50-289-4173