Learn More
Cayman Chemical THUNDRPhosphop70 S6KT38 480Wel
SDP
Supplier:  Cayman Chemical 500274480 WELL
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Phospho-p70 S6K (T389) in cell lysates
Catalog No. 50-289-6109