Learn More
Cayman Chemical THUNDRTotal CREB TRFRET C 2400
SDP
Supplier:  Cayman Chemical 5002442400 WELL
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Total CREB in cell lysates
Catalog No. 50-289-5409