Learn More
Cayman Chemical THUNDRTotal GAPDH TRFRET 2400
SDP
Supplier:  Cayman Chemical 5002492400 WELL
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Total GAPDH in cell lysates
Catalog No. 50-289-5899