Learn More
Cayman Chemical THUNDRTotal GAPDH TRFRET 4800
SDP
Supplier:  Cayman Chemical 5002494800 WELL
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Total GAPDH in cell lysates
Catalog No. 50-289-4159
Only null left
Add to Cart
Request a Quote