Learn More
Cayman Chemical THUNDRTotal GSK3 TRFRET 4800
SDP
Supplier:  Cayman Chemical 5002524800 WELL
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Total GSK3β in cell lysates
Catalog No. 50-289-3231
Only null left
Add to Cart
Request a Quote