Learn More
Cayman Chemical THUNDRTotal MEK1 TRFRET C 2400
SDP
Supplier:  Cayman Chemical 5002592400 WELL
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Total MEK1 in cell lysates
Catalog No. 50-289-5614