Learn More
Cayman Chemical THUNDRTotal MEK1 TRFRET C 9600
SDP
Supplier:  Cayman Chemical 5002599600 WELL
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Total MEK1 in cell lysates
Catalog No. 50-289-5265
Add to Cart
Request a Quote