Learn More
Cayman Chemical THUNDRTotal RPS6 TRFRET C 480
SDP
Supplier:  Cayman Chemical 500279480 WELL
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Total Ribosomal Protein S6 (both phosphorylated and non-phosphorylated RPS6) in cell lysates.
Catalog No. 50-289-4025