Learn More
Cayman Chemical THUNDRTotal SLP76 TRFRET 96Wel
SDP
Supplier:  Cayman Chemical 50022396 WELL
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Total SLP-76 (both phosphorylated and non-phosphorylated) in cell lysates
Catalog No. 50-289-5612