Learn More
Cayman Chemical THUNDRTotal STAT6 TRFRET 2400
SDP
Supplier:  Cayman Chemical 5002872400 WELL
Encompass_Preferred
Sandwich TR-FRET immunoassay for semi-quantitative detection of Total STAT6 (both phosphorylated and non-phosphorylated) in cell lysates.
Catalog No. 50-289-3423