Learn More
Cayman Chemical ThymIdIn-5-O- a -methylen 2mg
SDP
Supplier:  Cayman Chemical 385712 MG
Encompass_Preferred
A derivative of TDP and an inhibitor of thymidine kinase (Ki = 23 µM); has been used as a precursor in the synthesis of α,β-methylene-2’-deoxythymidine 5’-triphosphate
Catalog No. 50-291-9122