Learn More
Medchemexpress LLC Thymidine-13C | 478511-06-7 | 243.22 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN1150S21MG
Encompass_Preferred
Thymidine-13C is the 13C labeled Thymidine. It is a specific precursor of deoxyribonucleic acid, used as a cell synchronizing agent. As a DNA synthesis inhibitor, it can arrest cells at the G1/S boundary, prior to DNA replication. This product is for research use only and not sold to patients.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-15723