Learn More
Medchemexpress LLC Thymidine-15N2 | 50-89-5 | MFCD01073532 | 244.22 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN1150S91MG
Encompass_Preferred
Thymidine-15N2 is the 15N labeled Thymidine. Thymidine, a specific precursor of deoxyribonucleic acid, is used as a cell synchronizing agent. Thymidine is a DNA synthesis inhibitor that can arrest cell at G1/S boundary, prior to DNA replication.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process.
  • Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-16309