Learn More
Selleck Chemical LLC Thymidine 25mg 50-89-5 Deoxythymidine, 2'-Deoxythymidine, 5-Methyldeoxyuridine
SDP
Supplier:  Selleck Chemical LLC S480325MG
Encompass_Preferred
Thymidine (Deoxythymidine, 2'-Deoxythymidine, 5-Methyldeoxyuridine) is a pyrimidine nucleoside that is composed of the pyrimidine base thymine attached to the sugar deoxyribose. As a constituent of DNA, thymidine pairs with adenine in the DNA double helix. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8124