Learn More
Medchemexpress LLC Thymine-d4-1 | 156054-85-2 | 99.5% | 130.14 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW010450S4100MG
Encompass_Preferred
Thymine-d4-1 is a deuterium-labeled version of Thymine, one of the four nucleobases found in the nucleic acid of DNA. It can serve as a target for 5-fluorouracil (5-FU) in cancer treatment, with a Km of 2.3 μM. Deuterium substitution has shown potential in affecting the pharmacokinetic and metabolic profiles of drugs.
  • Application as a tracer
  • Internal standard for quantitative analysis
  • Stable isotope
Catalog No. 50-003-66487