Learn More
Medchemexpress LLC Thymine-d4-1 | 156054-85-2 | 99.5% | 130.14 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYW010450S4
Encompass_Preferred
Thymine-d4-1 is deuterium labeled Thymine, one of the four nucleobases in the nucleic acid of DNA. It can be a target for actions of 5-fluorouracil (5-FU) in cancer treatment, with a Km of 2.3 μM. This compound is a stable heavy isotope incorporated into drug molecules as tracers for quantitation during drug development. Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-66727