Learn More
Medchemexpress LLC Thyminose (deoxyribose) | 533-67-5 | 99.5% | C5H10O4 | 1 G
SDP
Supplier:  Medchemexpress LLC HY779561G
Encompass_Preferred
Thyminose is a natural 2-deoxyaldopentose, obtained by hydrolyzing nucleosides of thym nucleic acid. It provides positive reactions with Kilian's and Schiff's reagents and can consume the theoretical amount of iodine in Willstätter and Schudel oxidation methods. This product is for research use only.
  • Natural product
  • 2-deoxyaldopentose
  • Reacts positively with Kilian's reagent
  • Reacts positively with Schiff's reagent
  • Can consume theoretical amount of iodine in Willstätter and Schudel oxidation methods
  • Endogenous metabolite
  • Human gut microbiota metabolite
Catalog No. 50-004-35030