Learn More
Medchemexpress LLC HY-N6810 500mg Medchemexpress, Thymol CAS:89-83-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN6810500MG
Encompass_Preferred
Medchemexpress, HY-N6810 500mg Thymol CAS:89-83-8 Thymol is the main monoterpene phenol occurring in essential oils isolated from plants belonging to the Lamiaceae family, and other plants such as those belonging to the Verbenaceae, Scrophulariaceae, Ranunculaceae and Apiaceae families. Thymol has antioxidant, anti-inflammatory, antibacterial and antifungal effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2965