Learn More
Medchemexpress LLC Thymol blue | 76-61-9 | 1 G
SDP
Supplier:  Medchemexpress LLC HYD02191G
Encompass_Preferred
Thymol blue is a reversible pH indicator that responds to the pH of the solution through structural changes of protonation and deprotonation. It is red under acidic conditions and blue under alkaline conditions. This indicator can be used for quantitative detection through absorption peak shifts and selectively responds to pH changes, transitioning from red to yellow at pH 1.2-2.8 and from yellow to blue at pH 8.0-9.6.
  • Responds to pH of solution through structural changes of protonation and deprotonation.
  • Red under acidic conditions and blue under alkaline conditions.
  • Achieves quantitative detection through absorption peak shift (435 nm/596 nm).
  • Selectively responds to pH changes: red to yellow at pH 1.2-2.8, yellow to blue at pH 8.0-9.6.
  • Can be fixed in a silica gel matrix for in-situ pH measurement in marine environments.
  • Useful for acid-base monitoring in the biomedical field.
Catalog No. 50-004-11058