Learn More
Hach Company Thymol Blue Indicator Solution, 100 mL MDB
Supplier:  Hach Company 25732
Encompass
Thymol Blue Indicator Solution, Standardized for Colorimetric pH determination. pH Range: 7.8 - 10.0. 100 mL Marked Dropping Bottle
Catalog No. 50-199-4220