Learn More
Hach Company Thymolphthalein Indicator Solution, 1 g/L, 15 mL SCDB
Supplier:  Hach Company 2185336
Encompass
pH indicator: Visual transition interval: pH 8.8 (colorless)-pH 10.5 (blue). 15 mL Self-Contained Dropping Bottle. Replacement reagent for FM-1 test kit (2183100).
Catalog No. 50-199-5527