Learn More
Sigma Aldrich Fine Chemicals Biosciences Thymolphthalein ACS reagen50G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 11455350G
Encompass
Thymolphthalein (5AE 5AEAE-Diisopropyl-2AE 2AEAE-dimethylphenolphthalein) is an acid-base indicator which is colorless in SOLUTIONons with a pH below 8.5 and blue in SOLUTIONons with a pH above 9.5. Thymolphthalein has been used as an indicator in periodate-thiosulfate reaction.
Catalog No. 50-289-8798