Learn More
TARGETMOL CHEMICALS INC THYMOPENTIN 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T259810MG
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Synthetic pentapeptide corresponding to the amino acids 32-36 of thymopoietin and exhibiting the full biological activity of the natural hormone. Thymopentin (TP5) is an immunomodulator which has been studied for possible use in the treatment of rheumatoid arthritis AIDS and other primary immunodeficiencies. purity: 99%
Catalog No. 50-258-0731