Learn More
Cayman Chemical Thymopentn 5mg
SDP
Supplier:  Cayman Chemical 301195 MG
Encompass_Preferred
A pentapeptide fragment of thymopoietin; induces prothymocyte differentiation in isolated mouse splenocytes at 0.018-18 µM; increases proliferation of, and phytohemagglutinin-stimulated IL-2 production in, isolated human PBLs at 1 UG/ml; reduces the number of splenic autologous rosette-forming cells in athymic mice at 0.02-10 mg/kg; adoptive cell transfer of splenocytes isolated from tumor-bearing mice administered thymopentin (100 ng/animal) reduces recipient tumor growth in a murine Lewis lung carcinoma model
Catalog No. 50-298-8163