Learn More
CPC Scientific H-Arg-Lys-Asp-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific THYM003A
Encompass
TP-4 and TP-3; shortest active thymopoietin II fragments described that exhibit potent immunoregulatory properties in vitro and in vivo.SEQUENCE: H-Arg-Lys-Asp-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: RKDMOLECULAR FORMULA: C16H31N7O6MOLECULAR WEIGHT: 417.47STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [85465-82-3]SYNONYMS: RESEARCH AREA: ImmunomodulationREFERENCES:L. Kisfaludy et al., Hoppe-Seyler's Z. Physiol. Chem., 364, 933 (1983)L. Dnes et al., Drugs Exptl. Clin. Res. , 13, 279 (1987)O. Nyki et al., Peptides 1990, Proceedings of the 21st European Peptide Symposium, Platja d'Aro, E.Giralt and D.Andreu, edsSKU(s): THYM-003A
Catalog No. 50-211-0575