Learn More
Medchemexpress LLC Thymoquinone | 490-91-5 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYD080310MM/1ML
Encompass_Preferred
Thymoquinone is an orally active natural product isolated from N. sativa. It down-regulates the VEGFR2-PI3K-Akt pathway and exhibits a wide range of beneficial activities, including antioxidant, anti-inflammatory, and anticancer effects. This compound is valuable for research into various conditions such as Alzheimer's disease, cancer, cardiovascular disease, infectious disease, and inflammation.
  • Orally active natural product
  • Isolated from N. sativa
  • Down-regulates VEGFR2-PI3K-Akt pathway
  • Exhibits antioxidant and anti-inflammatory properties
  • Provides anticancer, antiviral, and antifungal activity
  • Offers anticonvulsant and antiangiogenic effects
  • Demonstrates hepatoprotective properties
  • Suitable for studies on Alzheimer's disease, cancer, and cardiovascular disease
  • Applicable for research into infectious disease and inflammation
Catalog No. 50-004-10935