Learn More
Bioworld THYMOSIN-A1
Supplier:  Bioworld 220607141
Encompass_Preferred
Thymosin-alpha 1 Source: Description : Thymosin alpha-1 acetate, also known as thymalfasin has immunoregulatory properties enhancing immune functions. Thymosin a1 has a molecular formula of C129H215N33O55 a.a. sequence of Ac-Ser-Asp-...
Catalog No. 50-253-812