Learn More
Medchemexpress LLC Thyroglobulin 25mg | 9010-34-8 | 25MG
SDP
Supplier:  Medchemexpress LLC HY10790325MG
Encompass_Preferred
Thyroglobulin is a 660 kDa dimeric glycoprotein produced by the follicular cells of the thyroid and used entirely within the thyroid gland Thyroglobulin acts as a substrate for the synthesis of the thyroid hormones thyroxine (T4) and triiodothyronine (T3) as well as the storage of the inactive forms of thyroid hormone and iodine within the follicular lumen of a thyroid follicle[1
Catalog No. 50-002-42653