Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Leica Biosystems Thyroid Transcription Factor-1 (TTF-1) antibody, clone SPT24 - liquid concentrate and BOND RTU formats

Supplier:  Leica Biosystems TTF1LU

Encompass

Monoclonal antibody directed against Thyroid Transcription Factor-1 (TTF-1), provided for detection of TTF-1 expression in normal and neoplastic tissues by immunohistochemistry. Available in liquid concentrate and ready-to-use formats for manual or automated staining workflows.

  • Target: Thyroid Transcription Factor-1 (TTF-1).
  • Clone: SPT24.
  • Application: Immunohistochemistry (IHC) for detection in normal and neoplastic tissues.
  • Formats: Liquid concentrate and BOND ready-to-use (RTU).
  • Pack sizes: Small volume and larger reagent formats available, for example 1 ml and 7 ml.
  • Use case: Adjunct to conventional histopathology for antigen localization.

Catalog No. NC3874030


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.