Learn More
Arbor Assays Thyroxine (T4) ELISA Kit (5 Plate)
SDP
Supplier:  Arbor Assays K050H5
Encompass_Preferred
The DetectX Thyroxine (T4) ELISA Kits quantitatively measure T4 present in serum, EDTA plasma, tissue culture media samples and fecal extracts.
Catalog No. 50-227-8468