Learn More
Medchemexpress LLC HY-136351 5mg Medchemexpress, THZ-P1-2 CAS:2058075-45-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1363515MG
Encompass_Preferred
Medchemexpress, HY-136351 5mg THZ-P1-2 CAS:2058075-45-7 THZ-P1-2 is a first-in-class and selective PI5P4K inhibitor, with an IC50 of 190 nM for PI5P4Kα. THZ-P1-2 covalently targets cysteines on a disordered loop in PI5P4Kα/β/γ. THZ-P1-2 causes autophagy disruption and upregulates TFEB signaling. THZ-P1-2 displays anticancer activity in leukemia cell lines. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1438