Learn More
TARGETMOL CHEMICALS INC THZ1 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T366410MG
Encompass_Preferred
Also available in 5 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. THZ1 (CDK7 inhibitor) is a novel selective and potent covalent CDK7 inhibitor. purity: 99%
Catalog No. 50-257-6891