Learn More
Selleck Chemical LLC THZ1 2HCl
SDP
Supplier:  Selleck Chemical LLC S75491G
Encompass_Preferred
THZ1 is a covalent CDK7 inhibitor which has the unprecedented ability to target a remote cysteine residue located outside of the canonical kinase domain providing an unanticipated means of achieving selectivity for CDK7
Catalog No. 50-000-04768