Learn More
Selleck Chemical LLC THZ1 2HCl 5mg 1604810-83-4(freebase)
SDP
Supplier:  Selleck Chemical LLC S75495MG
Encompass_Preferred
THZ1 is a covalent CDK7 inhibitor which has the unprecedented ability to target a remote cysteine residue located outside of the canonical kinase domain, providing an unanticipated means of achieving selectivity for CDK7. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4932